missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DPPIV/CD26 Activity Assay Kit (Fluorometric)
Assay Kit (Fluorometric)
Brand: Novus Biologicals™ NBP3-24487
This item is not returnable.
View return policy
Specifications
Storage of components varies. See protocol for specific instructions. | |
DPP4 | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
Non-species specific |
1803 | |
96 Tests | |
Adaptive Immunity, Cellular Markers, GPCR, Immunology |