Learn More
Elma™ EC 40 Cleaner
Providing powerful cleaning power for bridges, crowns, dental and medical instruments made from: stainless-steel, ceramics, plastics, glass and rubber.
Brand:  Elma™ 5819300000
 View more versions of this product
Product Code. 10436402
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Removes fast and efficiently dental cements from bridges, crowns, dental and medical instruments made from: stainless-steel, ceramics, plastics, glass and rubber.
  • Removes dental cements including phosphate cements, metal oxides, lime and fluxing agents.
  • Not suitable for light metals (e.g. AI, Mg, Zn and corresponding alloys).
Specifications
Specifications
EC 40 Cleaner
Can
10 L
CE
For removes dental cement