Learn More
Elma™ EC 40 Cleaner
Providing powerful cleaning power for bridges, crowns, dental and medical instruments made from: stainless-steel, ceramics, plastics, glass and rubber.
18.45 € - 317.97 €
Specifications
Certifications/Compliance CE
View More Specs
Products 4
Product Code Brand Volume (Metric) Container Type Product Type Price Quantity & Availability  
Product Code Brand Volume (Metric) Container Type Product Type Price Quantity & Availability  
10180522
View Documents
Elma™
5819100000
1 L Bottle EC 40 Cleaner This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10475632
View Documents
Elma™
5813700000
1 L Bottle EC 25 Cleaner
18.45 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10436402
View Documents
Elma™
5819300000
10 L Can EC 40 Cleaner This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10303982
View Documents
Elma™
5819400000
25 L Can EC 40 Cleaner This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Removes fast and efficiently dental cements from bridges, crowns, dental and medical instruments made from: stainless-steel, ceramics, plastics, glass and rubber.
  • Removes dental cements including phosphate cements, metal oxides, lime and fluxing agents.
  • Not suitable for light metals (e.g. AI, Mg, Zn and corresponding alloys).
Specifications
Specifications
CE