Learn More
Marienfeld Superior™ Embedding Cassettes with Standard Perforation
Use Marienfeld Superior™ Embedding cassettes in histology to dehydrate and impregnate tissue sections by Paraffin.
Brand:  Marienfeld Superior™ 5252501
 View more versions of this product
Product Code. 10434723
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Made of high-quality, technical plastic (POM)
  • Resistant against the solvents usually used in laboratories
  • Different colors offer the possibility of color coding (for example for different users, priorities, etc.)
  • With one 45° inclined and 2 vertial marking areas
  • The roughness of the marking areas is optimized to allow labelling by different systems: pencil, permanent markers, inkjet or laser printers
  • Suitable for all common clamps
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark and batch number for comprehensive information and traceability
Specifications
Specifications
1 mm x 5 mm
Yes
Base and Upper Part Connected by a Hinge
POM
45°
500
Yellow
Universal