Learn More
Memmert™ Entry Port 23 mm for UF750, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF750F0
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position left side center/center
Specifications
UF750 Oven | |
Entry Port | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.