Learn More
Equilin, MedChemExpress
MedChemExpress Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.
Brand: Medchem Express HY-B1176
Description
Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.Specifications
Equilin | |
White | |
C18H20O2 | |
7-Dehydroestrone | |
C[C@]1([C@](CC2)([H])C3=CCC4=C(C=CC(O)=C4)[C@@]3([H])CC1)C2=O | |
268.35 | |
Research |
474-86-2 | |
Research | |
50 mg | |
DMSO : 100 mg/mL (372.65 mM; Need ultrasonic) | |
268.35 | |
99.0% | |
Powder |