missing translation for 'onlineSavingsMsg'
Learn More

Filaggrin Mouse, Clone: FLG/1561, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 2312-MSM1-P0

 View more versions of this product

Product Code. 15939987

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Filaggrin
Cytoplasmic
FLG/1561
Unconjugated
NP_002007
Mouse
26-45kDa (Processed) ;350kDa (Profilaggrin)
Protein A/G Purified
RUO
2312
IgG1
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
FLG
Recombinant human Filaggrin protein fragment (aa 998-1104)
U-251-MG cells or Skin
20 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions