missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin Mouse, Clone: FLG/1561, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 510.00 €
Specifications
Antigen | Filaggrin |
---|---|
Clone | FLG/1561 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Specifications
Filaggrin | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
2312 | |
Recombinant human Filaggrin protein fragment (aa 998-1104) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
26-45kDa (Processed) ;350kDa (Profilaggrin) |
FLG/1561 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_002007 | |
FLG | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic | |
U-251-MG cells or Skin |