Learn More
Heparan Sulfate, MedChemExpress
MedChemExpress Heparan sulfate, a complex and linear polysaccharide, exists as part of glycoproteins named heparan sulfate proteoglycans, which are expressed abundantly on the cell surface and in the extracellular matrix.
Brand: Medchem Express HY-101916
Description
Heparan sulfate, a complex and linear polysaccharide, exists as part of glycoproteins named heparan sulfate proteoglycans, which are expressed abundantly on the cell surface and in the extracellular matrix.Specifications
Heparan Sulfate | |
9050-30-0 | |
25 mg | |
C12H19NO20S3 (monomer) | |
H2O : 100 mg/mL (Need ultrasonic) | |
Research |
Solid | |
White | |
Research | |
Cancer-Kinase/protease | |
O=S(OCC1O[C@@H](OC)[C@H](NS(=O)(O)=O)[C@@H](OS(=O)(O)=O)[C@H]1O[C@@H]2OC(C(O)=O)[C@H](O[C@@H]3OC(COS(=O)(O)=O)[C@H](O[C@@H]4OC(C(O)=O)[C@H](OC)[C@H](O)[C@H]4OS(=O)(O)=O)[C@H](O)[C@H]3NS(=O)(O)=O)[C@H](O)[C@H]2O)(O)=O.[n] |