missing translation for 'onlineSavingsMsg'
Learn More

Heparan Sulfate, MedChemExpress

MedChemExpress Heparan sulfate, a complex and linear polysaccharide, exists as part of glycoproteins named heparan sulfate proteoglycans, which are expressed abundantly on the cell surface and in the extracellular matrix.

Encompass_Preferred

180.90 € - 544.05 €

Specifications

Chemical Name or Material Heparan Sulfate
Physical Form Solid
CAS 9050-30-0
Color White
Purity Grade Notes Research
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16456367
Encompass_Preferred
View Documents
Medchem Express
HY-101916
5 mg
180.90 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16452806
Encompass_Preferred
View Documents
Medchem Express
HY-101916
25 mg
544.05 €
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16433638
Encompass_Preferred
View Documents
Medchem Express
HY-101916
10 mg
279.45 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Heparan sulfate, a complex and linear polysaccharide, exists as part of glycoproteins named heparan sulfate proteoglycans, which are expressed abundantly on the cell surface and in the extracellular matrix.
Specifications

Specifications

Heparan Sulfate
9050-30-0
Research
Cancer-Kinase/protease
O=S(OCC1O[C@@H](OC)[C@H](NS(=O)(O)=O)[C@@H](OS(=O)(O)=O)[C@H]1O[C@@H]2OC(C(O)=O)[C@H](O[C@@H]3OC(COS(=O)(O)=O)[C@H](O[C@@H]4OC(C(O)=O)[C@H](OC)[C@H](O)[C@H]4OS(=O)(O)=O)[C@H](O)[C@H]3NS(=O)(O)=O)[C@H](O)[C@H]2O)(O)=O.[n]
Solid
White
C12H19NO20S3 (monomer)
H2O : 100 mg/mL (Need ultrasonic)
Research
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Heparan Sulfate, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.