missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ErbB2 (HER-2) Fc Chimera Recombinant Protein

Recombinant Protein

342.00 € - 2210.00 €

Specifications

Accession Number P04626
Concentration 0.83 mg/mL
For Use With (Application) ELISA, Functional Assay
Format Lyophilized
Formulation PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17946331
View Documents
Invitrogen™
RP88054
100 μg
342.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
17916441
View Documents
Invitrogen™
RP88055
1 mg
2210.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1695 μL) be added to the vial to prepare a stock solution of 0.83 mg/mL. Concentration is measured by UV.

ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human ErbB2 (HER-2) Fc Chimera Recombinant Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.