Learn More
Gibco™ Human FGF-8a Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
Brand:  Gibco™ 100-25A-01M
 View more versions of this product
Product Code. 30087756
  • 3915.00 € / Pack of 2
Please to purchase this item. Need a web account? Register with us today!
Explore more special offers
Description
Description
FGF-8 is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8a, FGF-8b, FGF-8e and FGF-8f. The human and murine FGF-8a and FGF-8b are identical, unlike human and mouse FGF-8e and FGF-8f, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P55075
Protein with no preservative
21.2
2 x 500 μg
-20°C
<1 EU/μg
Human
Recombinant
Lyophilized
Functional Assay
2253
Human FGF-8 A
CHO cells
RUO
FGF8
Unconjugated
QHVREQSLVT DQLSRRLIRT YQLYSRTSGK HVQVLANKRI NAMAEDGDPF AKLIVETDTF GSRVRVRGAE TGLYICMNKK GKLIAKSNGK GKDCVFTEIV LENNYTALQN AKYEGWYMAF TRKGRPRKGS KTRQHQREVH FMKRLPRGHH TTEQSLRFEF LNYPPFTRSL RGSQRTWAPE PR
≥ 95% by SDS-PAGE gel and HPLC analyses.