missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-8a Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
81.50 € - 3960.35 €
Specifications
Accession Number | P55075 |
---|---|
For Use With (Application) | Functional Assay |
Gene ID (Entrez) | 2253 |
Molecular Weight (g/mol) | 21.2 |
Name | Human FGF-8 A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17810703
|
Gibco™
100-25A-100UG |
100 μg |
756.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17820293
|
Gibco™
100-25A-1MG |
2 x 500 μg | N/A | ||||||
30087756
|
Gibco™
100-25A-01M |
2 x 500 μg |
3915.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17890493
|
Gibco™
100-25A-250UG |
250 μg |
1374.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17810293
|
Gibco™
100-25A-25UG |
25 μg |
198.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17891753
|
Gibco™
100-25A-500UG |
500 μg |
2360.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880493
|
Gibco™
100-25A-5UG |
5 μg |
81.50 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-8 is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8a, FGF-8b, FGF-8e and FGF-8f. The human and murine FGF-8a and FGF-8b are identical, unlike human and mouse FGF-8e and FGF-8f, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.Specifications
P55075 | |
2253 | |
Human FGF-8 A | |
-20°C | |
<1 EU/μg | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
21.2 | |
CHO cells | |
RUO | |
FGF8 | |
Unconjugated | |
QHVREQSLVT DQLSRRLIRT YQLYSRTSGK HVQVLANKRI NAMAEDGDPF AKLIVETDTF GSRVRVRGAE TGLYICMNKK GKLIAKSNGK GKDCVFTEIV LENNYTALQN AKYEGWYMAF TRKGRPRKGS KTRQHQREVH FMKRLPRGHH TTEQSLRFEF LNYPPFTRSL RGSQRTWAPE PR | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title