missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GDNF, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ AF-450-10-500UG
This item is not returnable.
View return policy
Description
GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.Specifications
P39905 | |
protein with no preservative | |
30.4 | |
500 μg | |
-20°C | |
<0.1 EU/μg | |
GDNF | |
Unconjugated | |
MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
2668 | |
Human GDNF, Animal-Free | |
E. coli | |
RUO | |
AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
Human | |
Recombinant | |
Lyophilized |