missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human GDNF, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
220.00 € - 5630.00 €
Specifications
Accession Number | P39905 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 2668 |
Molecular Weight (g/mol) | 30.4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17873753
|
Gibco™
AF-450-10-100UG |
100 μg |
1170.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17803453
|
Gibco™
AF-450-10-10UG |
10 μg |
220.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17853053
|
Gibco™
AF-450-10-1MG |
1 mg |
5630.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17803393
|
Gibco™
AF-450-10-500UG |
500 μg |
3375.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.Specifications
P39905 | |
protein with no preservative | |
30.4 | |
E. coli | |
RUO | |
AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
2668 | |
Human GDNF, Animal-Free | |
-20°C | |
<0.1 EU/μg | |
GDNF | |
Unconjugated | |
MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |