missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG4 Mouse, Clone: IGHG4/1345, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3503-MSM1-P1
This item is not returnable.
View return policy
Specifications
IgG4 | |
Cytoplasmic | |
IGHG4/1345 | |
Unconjugated | |
IGHG4 | |
Recombinant human full-length IGHG4 | |
Tonsil | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Mouse | |
150kDa | |
Protein A/G Purified | |
RUO | |
3503 | |
IgG1 |