missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG4 Mouse, Clone: IGHG4/1345, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 510.00 €
Specifications
Antigen | IgG4 |
---|---|
Clone | IGHG4/1345 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Specifications
IgG4 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
IGHG4 | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic | |
Tonsil |
IGHG4/1345 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
3503 | |
Recombinant human full-length IGHG4 | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
150kDa |