missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inorganic Pyrophosphatase/PPA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP1-87788
This item is not returnable.
View return policy
Description
Inorganic Pyrophosphatase/PPA1 Polyclonal specifically detects Inorganic Pyrophosphatase/PPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Inorganic Pyrophosphatase/PPA1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:500 | |
diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PPA1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GFSTEERAAPFSLEYRVFLKNEKGQYISPFHDIPIYADKDVFHMVVEVPRWSNAK | |
0.1 mL | |
metabolism, Signal Transduction | |
5464 | |
Human, Mouse, Rat | |
IgG |
For Research Use Only