Learn More
DWK Life Sciences Kimble™ Kontes™ Tube For Potter-Elvehjem Tissue Grinder
A graduated tube with markings fused onto the glass. Range of tube sizes: Size 20 (4mL), Size 21 (10mL) and Size 22 (55mL).
Brand:  DWK Life Sciences 885752-0019
 View more versions of this product
Product Code. 10315362
  • 68.55 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Graduations are fused onto the glass
  • Clearance between pestle and tube
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
19
Tissue Grinder Tube
Potter-Elvehjem tissue grinder