missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Tube For Potter-Elvehjem Tissue Grinder

A graduated tube with markings fused onto the glass. Range of tube sizes: Size 20 (4mL), Size 21 (10mL) and Size 22 (55mL).

68.55 € - 120.00 €

Specifications

Product Type Tissue Grinder Tube
For Use With (Equipment) Potter-Elvehjem tissue grinder
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10633823
View Documents
DWK Life Sciences
885512-0023
23
101.00 €
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
10315362
View Documents
DWK Life Sciences
885752-0019
19
68.55 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
10355402
View Documents
DWK Life Sciences
885512-0020
20
120.00 €
Each
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
10613633
View Documents
DWK Life Sciences
885512-0021
21
68.55 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
 
Description

Description

  • Graduations are fused onto the glass
  • Clearance between pestle and tube
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications

Specifications

Tissue Grinder Tube
Potter-Elvehjem tissue grinder
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.