Learn More
DWK Life Sciences Kimble™ Kontes™ Tube For Potter-Elvehjem Tissue Grinder
A graduated tube with markings fused onto the glass. Range of tube sizes: Size 20 (4mL), Size 21 (10mL) and Size 22 (55mL).
Brand: DWK Life Sciences 885512-0021
This item is not returnable.
View return policy
Description
- Graduations are fused onto the glass
- Clearance between pestle and tube
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
21 | |
Tissue Grinder Tube | |
Potter-Elvehjem tissue grinder |