missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Homoserine, 99%, Thermo Scientific Chemicals
40.40 € - 383.00 €
Chemical Identifiers
CAS | 672-15-1 |
---|---|
Molecular Formula | C4H9NO3 |
Molecular Weight (g/mol) | 119.12 |
MDL Number | MFCD00063090 |
InChI Key | UKAUYVFTDYCKQA-VKHMYHEASA-N |
Synonym | l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo |
PubChem CID | 12647 |
ChEBI | CHEBI:15699 |
IUPAC Name | (2S)-2-amino-4-hydroxybutanoic acid |
SMILES | C(CO)C(C(=O)O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338805
|
Thermo Scientific Alfa Aesar
L12427.MD |
250 mg |
40.40 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318805
|
Thermo Scientific Alfa Aesar
L12427.03 |
1 g |
111.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328805
|
Thermo Scientific Alfa Aesar
L12427.06 |
5 g |
383.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.
Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Chemical Identifiers
672-15-1 | |
119.12 | |
UKAUYVFTDYCKQA-VKHMYHEASA-N | |
12647 | |
(2S)-2-amino-4-hydroxybutanoic acid |
C4H9NO3 | |
MFCD00063090 | |
l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo | |
CHEBI:15699 | |
C(CO)C(C(=O)O)N |
Specifications
∽203°C (decomposition) | |
C4H9NO3 | |
1721681 | |
l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo | |
UKAUYVFTDYCKQA-VKHMYHEASA-N | |
(2S)-2-amino-4-hydroxybutanoic acid | |
12647 | |
119.12 | |
−9° (c=2 in Water) |
672-15-1 | |
MFCD00063090 | |
14,4739 | |
Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol. | |
C(CO)C(C(=O)O)N | |
119.12 | |
CHEBI:15699 | |
99% | |
L-Homoserine |
Safety and Handling
missing translation for 'einecsNumber' : 211-590-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only