Learn More
Thermo Scientific™ LanthaScreen™ STAT3 GripTite™ Cell Line
The JAK/STAT3 signaling pathway is known to be activated by cytokines such as IL-6. In this pathway, binding of IL-6 to its cell-surface receptors results in the activation of JAKs, which in turn phosphoactivate STAT3 proteins at a specific tyrosine residue (Y705).
Brand:  Thermo Scientific™ K1488
Product Code. 10104643
  • 11390.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
The JAK/STAT3 signaling pathway is known to be activated by cytokines such as IL-6. In this pathway, binding of IL-6 to its cell-surface receptors results in the activation of JAKs, which in turn phosphoactivate STAT3 proteins at a specific tyrosine residue (Y705). LanthaScreen™ STAT3 GripTite™ is a human cell line that constitutively expresses a GFP-STAT3 fusion protein. The JAK/STAT signaling pathway is known to be functionally intact in this cell line; therefore, the GFP-STAT3 fusion protein serves as a substrate for IL-6-inducible phosphorylation. Using this cell line, a lytic immunoassay has been developed in which the phosphorylation state of GFP-STAT3 is detected in cell lysates using a terbium-labeled anti-pY705-STAT3 antibody in a time-resolved FRET (TR-FRET) readout.
The GFP-STAT3 DNA expression construct was transfected into the GripTite™ HEK 293 cell line using Lipofectamine™ 2000 transfection reagent, and the transfected cells were selected with blasticidin. This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as a sorting marker. The assay utilizing this cell line is validated for EC50 and Z'-factors under optimized conditions using IL-6 as a ligand for JAK-mediated GFP-STAT3 phosphorylation. This assay has also been tested under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance, and assay equilibration time. An alternate 'mix-and-read' assay format is also described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-6
Cell-based terbium phospho-specific activity (stable)
LanthaScreen™ STAT3 GripTite™ cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
STAT3
GripTite™, LanthaScreen™
End Point
STAT3
LanthaScreen™
IFN-alpha
Dividing Cells
Cytokines, Kinases
IL-6
1 Vial
GFP (EmGFP)
Dividing Cells
Kinases