Learn More
MP Biomedicals™ Lipase, MP Biomedicals™
Catalyze the hydrolysis and synthesis of the emulsified esters of glycerol and long chain fatty acids. Lipase, MP Biomedicals is a subclass of the esterases. It is used industrially for the resolution of chiral compounds and the trans-esterification produ
Brand: MP Biomedicals™ 0210218920
Description
Lipases catalyze the hydrolysis of esters in aqueous solutions and the synthesis of esters in non-aqueous solutions.They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).

Specifications
Cream | |
-20°C | |
20 g |
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel. | |
Porcine | |
Powder |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- lipase
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : lipase
Your input is important to us. Please complete this form to provide feedback related to the content on this product.