Learn More
WTW™ LR 925/01 2 Electrode Conductivity Cell
The conductivity measuring cells of the highest quality for decades. These are the basis for the new IDS conductivity measuring cells. The advantages of the IDS technology include the automatic transfer of the cell constant and therefore the exclusion of possible erroneous input.
Brand:  WTW™ 301720
858.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11745184
  • 882.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
2-electrodes, Graphite
0°C to 100°C
120 mm
1.5 m
12 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ LR 925/01 2 Electrode Conductivity Cell > 2 electrodes; Material: Stainless Steel; Cell Constant: 0.1cm; Cable Length: 1.5m