missing translation for 'onlineSavingsMsg'
Learn More

LRIG3 Antibody (814834) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB3495J

Product Code. 18610900

  • 623.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

LRIG3 Monoclonal antibody specifically detects LRIG3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRIG3
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
121227
Store at 4°C in the dark.
IgG2b
Western Blot, Flow Cytometry, CyTOF
814834
Western Blot, Flow Cytometry, CyTOF-ready
FLJ90440, KIAA3016, leucine-rich repeats and immunoglobulin-like domains 3, leucine-rich repeats and immunoglobulin-like domains protein 3, LIG-3, LIG3FLJ26573, LIRG3
Mouse myeloma cell line NS0-derived recombinant human LRIG3, Asp28-Thr807, Accession # Q6UXM1
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LRIG3 Antibody (814834) [Janelia Fluor™ 646], Novus Biologicals™ > 0.1 mL; Janelia Fluor 646

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.