missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LSD1 Activity/Inhibition Assay Kit (Fluorometric)
Assay Kit (Fluorometric)
Brand: Novus Biologicals™ KA1603
This item is not returnable.
View return policy
Specifications
Storage of components varies. See protocol for specific instructions. | |
KDM1A | |
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A |
23028 | |
1 Kit | |
DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics |