Description
- Double sole design
- Elasticated opening and tie fastening
- Ultrasonically welded seams
- Multi-layer barrier laminate
Specifications
Specifications
Size | Fits over Euro sizes 42-46 (UK 8-11) |
Material | Multi-layer Barrier Laminate Fabric |
Color | Green |
Closure Type | Elastic Opening and Ties |
Sole Type | Double Sole Design |
Certifications/Compliance (Europe) | Category I PPE, 89/686/EEC |
Packaging Quantity | 100/Pk |
Product Type | Boot Covers |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell Microchem™ 4000 Overboots >
Spot an opportunity for improvement?Share a Content Correction