missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ Pre-Printed Polypropylene Labels
These MiniMark™ Pre-Printed Polypropylene Labels are supplied with pre-printed red diamonds and are ready for you to print the required warning.
Brand: Brady™ 710554
This item is not returnable.
View return policy
Description
- Supplied with pre-printed red diamonds.
- Ready for you to print the required warning.
- Compliant to the latest regulations.
Specifications
RoHS compliant using EU Directive 2002/95/EC | |
Permanent Acrylic, Permanent | |
Gloss | |
Thermal Transfer | |
Hazard Communication Identification | |
-20°C | |
400 Pk. | |
-4°F to +212°F | |
0.0031 in. | |
55 x 85 mm | |
55 mm | |
85 mm |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | |
Red on White | |
Polypropylene | |
Labels | |
MiniMark Printer | |
400 Labels/Box | |
Diamond | |
-20°C to +100°C | |
0.079 mm | |
2.17 in. | |
3.35 in. |