missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ Pre-Printed Polypropylene Labels
These MiniMark™ Pre-Printed Polypropylene Labels are supplied with pre-printed red diamonds and are ready for you to print the required warning.
181.00 € - 499.00 €
Specifications
Description | Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances |
---|---|
Adhesive Type | Permanent Acrylic, Permanent |
Color | Red on White |
Finish | Gloss |
Material | Polypropylene |
Product Code | Brand | Description | HeightMetric | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | HeightMetric | Width (Metric) | Price | Quantity & Availability | |||
11799465
|
Brady™
710563 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 110 mm | N/A | |||||
11709475
|
Brady™
710564 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 110 mm |
339.00 €
1 box |
In Stock
Log in to see stock. |
||||
11719475
|
Brady™
710565 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 110 mm |
339.00 €
1 box |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||||
11729475
|
Brady™
710566 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 110 mm | N/A | |||||
11749475
|
Brady™
710568 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 110 mm | N/A | |||||
11739475
|
Brady™
710567 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 110 mm | N/A | |||||
11779455
|
Brady™
710575 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 155 mm |
499.00 €
1 box |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||||
11789455
|
Brady™
710576 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 155 mm |
499.00 €
1 box |
In Stock
Log in to see stock. |
||||
11799455
|
Brady™
710577 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 155 mm | N/A | |||||
11709465
|
Brady™
710578 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 155 mm | N/A | |||||
11719465
|
Brady™
710579 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 155 mm |
499.00 €
1 box |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||||
11729485
|
Brady™
710552 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 85 mm |
241.00 €
1 box |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||||
11719485
|
Brady™
710551 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 85 mm | N/A | |||||
11739485
|
Brady™
710553 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 85 mm |
241.00 €
1 box |
In Stock
Log in to see stock. |
||||
11749485
|
Brady™
710554 |
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | 85 mm | N/A | |||||
Description
- Supplied with pre-printed red diamonds.
- Ready for you to print the required warning.
- Compliant to the latest regulations.
Specifications
Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances | |
Red on White | |
Polypropylene | |
Labels | |
MiniMark Printer | |
Diamond | |
-20°C to +100°C | |
0.079 mm |
Permanent Acrylic, Permanent | |
Gloss | |
Thermal Transfer | |
Hazard Communication Identification | |
-20°C | |
-4°F to +212°F | |
0.0031 in. |