Learn More
Hach™ Molybdate 3 Reagent
For low and ultra-low range silica determination by the Heteropoly Blue method.
Brand:  Hach™ 199549
 View more versions of this product
Product Code. 10602834
  • 39.90 € / 500mL
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Molybdate
Molybdate determination
250
3 to 1,000μg/L SiO2
Reagent
Solution
Spectrophotometer, Pour-thru cell
500 mL
Heteropoly blue (Hach methods 8282 and 8186)