missing translation for 'onlineSavingsMsg'
Learn More

Hach™ Molybdate 3 Reagent

For low and ultra-low range silica determination by the Heteropoly Blue method.

39.90 €

Specifications

Analytes Molybdate
Format Solution
For Use With (Application) Molybdate determination
For Use With (Equipment) Spectrophotometer, Pour-thru cell
Range 3 to 1,000μg/L SiO2
View More Specs

Products 1
Product Code Brand Quantity No. of Tests Price Quantity & Availability  
Product Code Brand Quantity No. of Tests Price Quantity & Availability  
10602834
View Documents
Hach™
199549
500 mL 250
39.90 €
500mL
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Molybdate
Molybdate determination
3 to 1,000μg/L SiO2
Reagent
Solution
Spectrophotometer, Pour-thru cell
Heteropoly blue (Hach methods 8282 and 8186)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.