missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-aspartic acid, 98%, Thermo Scientific Chemicals
24.60 € - 66.30 €
Chemical Identifiers
CAS | 13726-67-5 |
---|---|
Molecular Formula | C9H15NO6 |
Molecular Weight (g/mol) | 233.22 |
MDL Number | MFCD00037279 |
InChI Key | KAJBMCZQVSQJDE-YFKPBYRVSA-N |
Synonym | boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid |
PubChem CID | 99718 |
IUPAC Name | (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid |
SMILES | CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358195
|
Thermo Scientific Alfa Aesar
L08498.03 |
1 g |
24.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368195
|
Thermo Scientific Alfa Aesar
L08498.06 |
5 g |
66.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13726-67-5 | |
233.22 | |
KAJBMCZQVSQJDE-YFKPBYRVSA-N | |
99718 | |
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O |
C9H15NO6 | |
MFCD00037279 | |
boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid |
Specifications
13726-67-5 | |
C9H15NO6 | |
1913973 | |
KAJBMCZQVSQJDE-YFKPBYRVSA-N | |
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O | |
233.22 | |
233.23 | |
N-Boc-L-aspartic acid |
115°C | |
MFCD00037279 | |
boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid | |
−5.5° (c=1 in Methanol) | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid | |
99718 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 237-294-7
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only