missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-D-aspartic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals
29.70 € - 210.00 €
Chemical Identifiers
CAS | 134098-70-7 |
---|---|
Molecular Formula | C23H25NO6 |
Molecular Weight (g/mol) | 411.454 |
MDL Number | MFCD00237011 |
InChI Key | VZXQYACYLGRQJU-LJQANCHMSA-N |
Synonym | fmoc-d-asp-otbu, fmoc-d-aspartic acid 1-tert-butyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid alpha-t-butyl ester, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp otbu, ambotzfaa1311, fmoc-d-aspartic acid-otbu, fmoc-d-asp-tert-butoxide, fmoc-d-aspartic acid alpha-t-butyl ester |
PubChem CID | 7019703 |
IUPAC Name | (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
SMILES | CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15417726
|
Thermo Scientific Alfa Aesar
H62680.MD |
250 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15479168
|
Thermo Scientific Alfa Aesar
H62680.03 |
1 g |
65.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15489168
|
Thermo Scientific Alfa Aesar
H62680.06 |
5 g |
210.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-D-aspartic acid 1-tert-butyl ester is an pharmaceutical intermediate.
Solubility
Soluble in DMF.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
134098-70-7 | |
411.454 | |
VZXQYACYLGRQJU-LJQANCHMSA-N | |
7019703 | |
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
C23H25NO6 | |
MFCD00237011 | |
fmoc-d-asp-otbu, fmoc-d-aspartic acid 1-tert-butyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid alpha-t-butyl ester, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp otbu, ambotzfaa1311, fmoc-d-aspartic acid-otbu, fmoc-d-asp-tert-butoxide, fmoc-d-aspartic acid alpha-t-butyl ester | |
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
Specifications
134098-70-7 | |
MFCD00237011 | |
Soluble in DMF. | |
CC(C)(C)OC(=O)C(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
411.454 | |
411.45 | |
N-Fmoc-D-aspartic acid 1-tert-butyl ester |
C23H25NO6 | |
fmoc-d-asp-otbu, fmoc-d-aspartic acid 1-tert-butyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid alpha-t-butyl ester, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp otbu, ambotzfaa1311, fmoc-d-aspartic acid-otbu, fmoc-d-asp-tert-butoxide, fmoc-d-aspartic acid alpha-t-butyl ester | |
VZXQYACYLGRQJU-LJQANCHMSA-N | |
(3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
7019703 | |
95% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only