Learn More
NALP6 Antibody (920631) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9145J
Description
NALP6 Monoclonal antibody specifically detects NALP6 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
NALP6 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
920631 | |
Flow Cytometry, CyTOF-ready | |
Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
E.coli-derived recombinant human NLRP6/NALP6, Lys35-Arg193, Accession # P59044 | |
0.1 mL | |
Immunology, Inflammation | |
171389 | |
Store at 4°C in the dark. | |
IgG2b |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.