Learn More
Deutsch Neumann™ Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Natural Rubber Stoppers are suitable for general use.
Brand: Deutsch Neumann™ 1011121
This item is not returnable.
View return policy
Description
- Acc. to DIN 12 871
- Durometer 40 Shore A
- Available in a wide range of sizes
- Available in either grey or red color
Specifications
1 hole, 5 mm dia. | |
40 Shore A | |
1/Pk. | |
21 mm | |
Natural Rubber |
5mm | |
30mm | |
Gray | |
27 mm | |
Stopper |