Learn More
Deutsch Neumann™ Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Natural Rubber Stoppers are suitable for general use.
0.20 € - 21.60 €
Specifications
Material Natural Rubber
Hardness 40 Shore A
View More Specs
Products 10
Product Code Brand Diameter (Metric) Bottom Color Description Price Quantity & Availability  
Product Code Brand Diameter (Metric) Bottom Color Description Price Quantity & Availability  
10475801
View Documents
Deutsch Neumann™
1010179
79 mm Gray Solid
13.80 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
10068630
View Documents
Deutsch Neumann™
1011118
18 mm Gray 1 hole, 4 mm dia. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10199930
View Documents
Deutsch Neumann™
1010187
87 mm Gray Solid
16.20 €
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
10672791
View Documents
Deutsch Neumann™
1011112
12.5 mm Gray 1 hole, 3 mm dia. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10199980
View Documents
Deutsch Neumann™
1011121
21 mm Gray 1 hole, 5 mm dia. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10221731
View Documents
Deutsch Neumann™
1011123
23 mm Gray 1 hole, 5 mm dia.
0.85 €
Each
In Stock
to see stock.
Edge
Add to basket
10088640
View Documents
Deutsch Neumann™
1011126
26 mm Gray 1 hole, 6 mm dia. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10672981
View Documents
Deutsch Neumann™
1011129
29 mm Gray 1 hole, 6 mm dia. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10534173
View Documents
Deutsch Neumann™
1010294
94 mm Red Solid
21.60 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
10711421
View Documents
Deutsch Neumann™
1010210
10.5 mm Red Solid
0.25 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
  • Available in either grey or red color
Specifications
Specifications
Natural Rubber
40 Shore A