Learn More
Deutsch Neumann™ Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Natural Rubber Stoppers are suitable for general use.
Brand:  Deutsch Neumann™ 1010210
 View more versions of this product
Product Code. 10711421
  • 0.25 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
  • Available in either grey or red color
Specifications
Specifications
Solid
20mm
Red
14.5 mm
Tapered Stopper
40 Shore A
1/Pk.
10.5 mm
Natural Rubber