Learn More
Deutsch Neumann™ Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Natural Rubber Stoppers are suitable for general use.
Brand:  Deutsch Neumann™ 1011112
 View more versions of this product
Product Code. 10672791
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
  • Available in either grey or red color
Specifications
Specifications
1 hole, 3 mm dia.
40 Shore A
1/Pk.
12.5 mm
Natural Rubber
3mm
20mm
Gray
16.5 mm
Stopper