missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NHLH1 (Human) Recombinant Protein

Human NHLH1 full-length ORF ( AAH13789, 1 a.a. - 133 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004807-P01.25ug

 View more versions of this product

Product Code. 16148381

  • 508.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sequence: MMLNSDTMELDLPPTHSETESGFSDCGGGAGPDGAGPGGPGGGQARGPEPGEPGRKDLQHLSREERRRRRRATAKYRTAHATRERIRVEAFNLAFAELRKLLPTLPPDKKLSKIEILRLAICYISYLNHVLDV
Specifications

Specifications

AAH13789
nescient helix loop helix 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NHLH1
GST
4807
Wheat germ expression system
25 μg
HEN1, NSCL, NSCL1, bHLHa35
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions