missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NME1 (Human) Recombinant Protein
Human NME1 full-length ORF ( AAH00293, 1 a.a. - 152 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004830-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MANCERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVGLKFMQASEDLLKEHYVDLKDRPFFAGLVKYMHSGPVVAMVWEGLNVVKTGRVMLGETNPADSKPGTIRGDFCIQVGRNIIHGSDSVESAEKEIGLWFHPEELVDYTSCAQNWIYE
Specifications
AAH00293 | |
non-metastatic cells 1, protein (NM23A) expressed in | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NME1 | |
GST |
4830 | |
Wheat germ expression system | |
25 μg | |
AWD, GAAD, NB, NBS, NDPK-A, NDPKA, NM23, NM23-H1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |