missing translation for 'onlineSavingsMsg'
Learn More

NSE gamma Mouse, Clone: ENO2/1375, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 2026-MSM1-P1

 View more versions of this product

Product Code. 15949947

  • 510.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NSE gamma
Cytoplasmic
ENO2/1375
Unconjugated
NP_001966
Mouse
50kDa
Protein A/G Purified
RUO
2026
IgG2b
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
ENO2
A synthetic peptide corresponding to aa416-433 of human NSE gamma
HepG2, SH-SY-5Y, HeLa or Y79 cells. Pancreas, Cerebellum or Pheochromocytoma.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions