Learn More
3M™ Oil Sorbent Boom
A double boom with two, 4 inch scrims encased tubes of particulate sorbent material, secured by a UV resistant netting.
Brand: 3M™ 7000088226
This item is not returnable.
View return policy
Description
- Used to cover large spills, or as a precaution in areas where large spills may occur
- Ideal for absorbing oil from areas where water may also be present
- Link sorbent booms together by built-in connectors to deploy over large areas
- UV-resistant to prevent deterioration and disintegration
- Designed for long-term use in ditches, streams, rivers, lakes, docks, and piers
- Cost-effective as oil, not water, is recovered
- Can be used outside or in-plant
Specifications
Sorbent Boom | |
200 mm | |
5 m |
Brown | |
Workplace Safety, Spill Prevention | |
T-270 GA |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Oil Sorbent Boom > 5 m, 200 mm
Spot an opportunity for improvement?Share a Content Correction