Learn More
Amarell™ Partial Immersion Eco-friendly Thermometer
Brand:  Amarell™ G11388
 View more versions of this product
Product Code. 10404862
  • 28.00 € / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Green Liquid
Partial Immersion
Partial Immersion Thermometer
-10°C
1°C
General use
300 mm
110°C
-10°C to +110°C
76 mm