Learn More
Amarell™ Partial Immersion Eco-friendly Thermometer
Brand: Amarell™ G11408
This item is not returnable.
View return policy
Specifications
Green Liquid | |
Partial Immersion | |
Partial Immersion Thermometer | |
-10°C | |
2°C |
General use | |
300 mm | |
360°C | |
-10°C to +360° C | |
76 mm |