missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Pear-Shaped Separatory Funnel

Separate (partition) the components of a mixture into two immiscible solvent phases of different densities. Karl Hecht™ Pear-Shaped Separatory Funnel is constructed of Duran™ and high-density polyethylene.

Brand:  Karl Hecht™ 42970030

 View more versions of this product

Product Code. 10043941

  • 52.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Duran™ construction.
  • NS stopcock.
  • Interchangeable plug.
  • Reliable retaining device.
  • NS plastic stopper (high-density polyethylene, acid and alkali resistant).
  • Reinforced socket and neck.
  • marked with capacity and NS size.
  • In cardboard boxes of 2 pieces.
Specifications

Specifications

2.5mm
Borosilicate Glass (3.3)
No
19/26
High-Density Polyethylene
Funnel
100 mL
DIN ISO 4800
Interchangeable NS plug of solid glass
NS, 2.5mm bore
19
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.