Learn More
Karl Hecht™ Pear-Shaped Separatory Funnel
Separate (partition) the components of a mixture into two immiscible solvent phases of different densities. Karl Hecht™ Pear-Shaped Separatory Funnel is constructed of Duran™ and high-density polyethylene.
Brand:  Karl Hecht™ 42971066
 View more versions of this product
Product Code. 10469231
  • 118.00 € / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Duran™ construction.
  • NS stopcock.
  • Interchangeable plug.
  • Reliable retaining device.
  • NS plastic stopper (high-density polyethylene, acid and alkali resistant).
  • Reinforced socket and neck.
  • marked with capacity and NS size.
  • In cardboard boxes of 2 pieces.
Specifications
Specifications
1000 mL
No
NS, 6.0mm bore
29
Borosilicate Glass (3.3)
Interchangeable NS plug of PTFE
High-Density Polyethylene
Funnel