Learn More
Reitenspiess™ Pipe Cleaning Brush
High-quality brush with bleached bristles and a galvanized wire stem/handle.
Brand:  Reitenspiess™ 31300534
 View more versions of this product
Product Code. 10637932
  • 24.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Long, thin, tube-like shape for deep reach and cleaning efficiency.
  • Bleached bristles.
  • Eyelet.
  • Galvanized wire handle/stem.
  • Capped end
Specifications
Specifications
Pipe Cleaner Brush
Eyelet
150 mm
Galvanized Wire
1000 mm
30 mm