Learn More
Masterflex™ Polyethylene Male Pipe Adapter, pack of 100
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30625-46
This item is not returnable.
View return policy
Description
- Polyethylene Male Pipe Adapter
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
3/8 in. NPT(M) x 1/2 in. ID Tubing | |
HDPE | |
1/2 in. | |
3/8 in. NPT(M) | |
Ethylene Oxide | |
Straight | |
Male Pipe Adapter |
Not Autoclavable | |
100 Fittings | |
1/2 in. | |
1/2 in. | |
3/8 in. NPT(M) | |
Translucent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ Polyethylene Male Pipe Adapter, pack of 100 > 3/8 in. NPT(M) x 1/2 in. ID Tubing
Spot an opportunity for improvement?Share a Content Correction