Learn More
Carl Roth™ Polypropylene PCR Workstation
Brand:  Carl Roth™ K818.1
Encompass
 View more versions of this product
Product Code. 10086661
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
Autoclavable
Color: Neon Blue
Polypropylene
PCR workstations
Lid
50 mm